Learn More
BRAND™ Laboratory Forceps
Manufactured from either PMP or POM for durability and resistance.
Brand: BRAND™ 113020
385.69 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- PMP forceps with pointed ends, autoclavable at 121°C, pack of 10
- POM forceps with round ends, glass-fiber reinforced, pack of 5
Specifications
Rounded | |
250 mm | |
5 |
POM | |
Forceps |
Product Content Correction
Product Title
BRAND™ Laboratory Forceps > Material: POM; Ends: Rounded