Learn More
Hirschmann™ labopette™ fix
Ergonomically shaped manual pipette, effortless operation and tip ejector.
Brand:  Hirschmann™ 9475509
 View more versions of this product
Product Code. 11692259
  • 3015.00 NOK / Each
Estimated Shipment: 21-07-2025
to see stock.
Explore more special offers
Description
  • Fixed volume setting
  • With tip ejector and recalibration possibility
  • Autoclavable
Specifications
Specifications
0.8%
Blue: 10-1000 μL
Autoclavable
Manual Pipette
Calibration device, individual number and weighing protocol
1 Each
ISO 8655
Tip ejector
1
500 μL
0.3%
Product Title
Hirschmann™ labopette™ fix > Volume (Metric) Range: 500 μL