Learn More
Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly with Standard Taper 19/38 Joints
A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring.
4775.00 NOK - 7455.00 NOK
Specifications
Material | Borosilicate Glass |
---|---|
Includes | Glass top and bottom, Silicone Rubber ring seal |
Product Type | Flask Kit |
Standard Taper Joint | 19/38 |
Weight (English) | 3 lb. |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15103035
|
Labconco™
7550000 |
25 mL |
4775.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
15113035
|
Labconco™
7550200 |
50 mL |
4880.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
15123035
|
Labconco™
7550400 |
100 mL |
4990.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
15153035
|
Labconco™
7550900 |
750 mL |
6920.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
15163035
|
Labconco™
7551000 |
1000 mL |
7455.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
15133035
|
Labconco™
7550600 |
250 mL |
5370.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
15143035
|
Labconco™
7550800 |
500 mL |
6920.00 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
Description
A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring
- Flasks should be filled no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is assured.
- Adapters are required to attach flasks to freeze dry valve ports (sold separately).
Specifications
Borosilicate Glass | |
Flask Kit | |
3 lb. | |
Joint | |
1.4 kg |
Glass top and bottom, Silicone Rubber ring seal | |
19/38 | |
Reusable | |
Flat |
Product Title
Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly with Standard Taper 19/38 Joints