Learn More
OHAUS™ Lab Lift
OHAUS™ Lab Lift provides secure support for holding glassware at elevated positions.
Brand: OHAUS™ 30400007
1731.42 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Lab Lift | |
Clamp, Support, Lab-Lift, CLR-LBLFTA010 | |
102 x 102 (L x W) Platform |
Aluminum | |
64 to 127 mm |
Product Content Correction
Product Title
OHAUS™ Lab Lift > Material: Aluminum; Platform Size: 102 x 102 (L x W)