missing translation for 'onlineSavingsMsg'
Learn More
L-Valine tert-butyl ester hydrochloride, 97% Product Code.: 15452648

L-Valine tert-butyl ester hydrochloride, 97%

Product Code. 15452648
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15452648

Brand: Thermo Scientific Alfa Aesar H56207.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13518-40-6 | C9H20ClNO2 | 209.71 g/mol

It is involved in the synthesis of enantiomerically pure amino acid ester isocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is involved in the synthesis of enantiomerically pure amino acid ester isocyanates.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13518-40-6
Molecular Formula C9H20ClNO2
Molecular Weight (g/mol) 209.71
MDL Number MFCD00034863
InChI Key AUIVQIHTTVPKFS-FJXQXJEOSA-N
Synonym h-val-otbu.hcl, l-valine tert-butyl ester hydrochloride, s-tert-butyl 2-amino-3-methylbutanoate hydrochloride, l-valine t-butyl ester hydrochloride, tert-butyl 2s-2-amino-3-methylbutanoate hydrochloride, l-valinetert-butylesterhydrochloride, tert-butyl l-valinate hydrochloride, valine, 1,1-dimethylethyl ester, hydrochloride, h-val-otbu hcl, h-val-obut.hcl
PubChem CID 12490702
IUPAC Name tert-butyl 2-amino-3-methylbutanoate hydrochloride
SMILES Cl.CC(C)[C@H](N)C(=O)OC(C)(C)C

Specifications

Beilstein 3693622
Quantity 5 g
Formula Weight 209.72
Optical Rotation +4.4° (c=2 in Water)
Percent Purity 97%
Chemical Name or Material L-Valine tert-butyl ester hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number81202377 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56207.06,H56207.03
Lot Number81200974 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56207.06,H56207.03
Lot NumberBD220551 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56207.06,H56207.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Valine tert-butyl ester hydrochloride, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.