missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine, Multi-Compendial, U.S.P., J.T. Baker™
2020.00 NOK - 12185.00 NOK
Chemical Identifiers
CAS | 72-18-4 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00064220 |
InChI Key | KZSNJWFQEVHDMF-BYPYZUCNSA-N |
Synonym | l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin |
PubChem CID | 6287 |
ChEBI | CHEBI:16414 |
IUPAC Name | (2S)-2-amino-3-methylbutanoic acid |
SMILES | CC(C)[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15507974
|
Avantor J.T.Baker
2095-05 |
100 g | Poly Bottle |
2020.00 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15517974
|
Avantor J.T.Baker
2095-06 |
1 kg | Poly Bottle |
12185.00 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
72-18-4 | |
117.15 | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
6287 | |
(2S)-2-amino-3-methylbutanoic acid |
C5H11NO2 | |
MFCD00064220 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CHEBI:16414 | |
CC(C)[C@H](N)C(O)=O |
Specifications
72-18-4 | |
MFCD00064220 | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
(2S)-2-amino-3-methylbutanoic acid | |
6287 | |
117.15 | |
Multi-Compendial/USP |
C5H11NO2 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CC(C)[C@H](N)C(O)=O | |
117.15 | |
CHEBI:16414 | |
1L = 1.23kg | |
L-Valine |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Valine, Multi-Compendial, U.S.P., J.T. Baker™