missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%
CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol
468.59 NOK - 1065.00 NOK
Chemical Identifiers
CAS | 6100-19-2 |
---|---|
Molecular Formula | C8H14K4O13 |
Molecular Weight (g/mol) | 474.58 |
MDL Number | MFCD00013065 |
InChI Key | DFVWEDQSAWCYPI-UHFFFAOYNA-N |
Synonym | potassium tartrate, dipotassium tartrate, argol, dipotassium 2,3-dihydroxybutanedioate, potassium tatrate, potassium tartarate, ksc492e6f |
PubChem CID | 6100505 |
IUPAC Name | bis(2,3-dihydroxybutanedioic acid) hydrate tetrapotassium |
SMILES | O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11925061
|
Thermo Scientific Acros
427991000 |
100 g | N/A | ||||||
11935061
|
Thermo Scientific Acros
427995000 |
500 g |
1065.00 NOK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6100-19-2 | |
474.58 | |
DFVWEDQSAWCYPI-UHFFFAOYNA-N | |
6100505 | |
O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O |
C8H14K4O13 | |
MFCD00013065 | |
potassium tartrate, dipotassium tartrate, argol, dipotassium 2,3-dihydroxybutanedioate, potassium tatrate, potassium tartarate, ksc492e6f | |
bis(2,3-dihydroxybutanedioic acid) hydrate tetrapotassium |
Specifications
6100-19-2 | |
C8H14K4O13 | |
15, 7796 | |
Solubility in water: 226g/L (20°C). | |
O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O | |
474.58 | |
226.27 | |
Plastic bottle |
97% | |
MFCD00013065 | |
potassium tartrate, dipotassium tartrate, argol, dipotassium 2,3-dihydroxybutanedioate, potassium tatrate, potassium tartarate, ksc492e6f | |
DFVWEDQSAWCYPI-UHFFFAOYNA-N | |
bis(2,3-dihydroxybutanedioic acid) hydrate tetrapotassium | |
6100505 | |
97% | |
L-(+)-Tartaric acid dipotassium salt |
Safety and Handling
Product Identifier
- L(+)-Tartaric acid dipotassium salt
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%