missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, ≥99%, MP Biomedicals™

371.00 NOK - 1203.10 NOK

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11416252
View Documents Promotion Details
MP Biomedicals
0210287305
5 g
371.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
11426252
View Documents Promotion Details
MP Biomedicals
0210287325
25 g
371.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
11436252
View Documents Promotion Details
MP Biomedicals
0210287380
100 g
1203.10 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

56-45-1
105.093
MTCFGRXMJLQNBG-REOHCLBHSA-N
5951
(2S)-2-amino-3-hydroxypropanoic acid
C3H7NO3
MFCD00064224
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
CHEBI:17115
C(C(C(=O)O)N)O

Specifications

56-45-1
≥99%
MFCD00064224
MTCFGRXMJLQNBG-REOHCLBHSA-N
(2S)-2-amino-3-hydroxypropanoic acid
5951
105.09
L-Serine
White
C3H7NO3
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
C(C(C(=O)O)N)O
105.093
CHEBI:17115
Free-flowing Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-274-3

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Product Certifications