Learn More
L(-)-Mannose, 99+%, Thermo Scientific Chemicals
620.00 NOK - 6220.00 NOK
Chemical Identifiers
CAS | 10030-80-5 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.16 |
MDL Number | MFCD00136021 |
InChI Key | GZCGUPFRVQAUEE-BXKVDMCESA-N |
Synonym | l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose |
PubChem CID | 82308 |
ChEBI | CHEBI:37681 |
IUPAC Name | (2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal |
SMILES | OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10184710
|
Thermo Scientific Acros
227592500 |
250 mg | Glass bottle |
620.00 NOK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10665842
|
Thermo Scientific Acros
227590010 |
1 g | Glass bottle |
1715.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10464155
|
Thermo Scientific Acros
227590050 |
5 g | Glass Bottle |
6220.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10030-80-5 | |
180.16 | |
GZCGUPFRVQAUEE-BXKVDMCESA-N | |
82308 | |
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal |
C6H12O6 | |
MFCD00136021 | |
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose | |
CHEBI:37681 | |
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O |
Specifications
10030-80-5 | |
White | |
0.5% max. (60° to 70°C) | |
99% min. (ELSD) (HPLC) | |
MFCD00136021 | |
− 14.70 | |
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether | |
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O | |
180.16 | |
CHEBI:37681 | |
99+% | |
L(-)-Mannose |
129.0°C to 131.0°C | |
0.1% max. | |
Authentic | |
C6H12O6 | |
−14.70° (20°C c=4,H2O,NH3) | |
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose | |
GZCGUPFRVQAUEE-BXKVDMCESA-N | |
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal | |
82308 | |
180.16 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 233-080-2
- L(-)-Mannose
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.