missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine Hydrochloride, 99+%, MP Biomedicals
Brand: MP Biomedicals 0219469791
Quantity | 1 kg |
---|
This item is not returnable.
View return policy
Description

Chemical Identifiers
657-27-2 | |
182.648 | |
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisylShow More | |
CHEBI:53633 | |
C(CCN)CC(C(=O)O)N.Cl |
C6H15ClN2O2 | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
69568 | |
(2S)-2,6-diaminohexanoic acid;hydrochloride |
Specifications
657-27-2 | |
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
C(CCN)CC(C(=O)O)N.Cl | |
182.648 | |
69568 | |
182.6 | |
Crystals |
C6H15ClN2O2 | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
(2S)-2,6-diaminohexanoic acid;hydrochloride | |
1 kg | |
CHEBI:53633 | |
Cell Culture | |
L-Lysine, 99+%, Hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Lysine Hydrochloride, 99+%, MP Biomedicals > 1kg
Spot an opportunity for improvement?Share a Content Correction