Learn More
L-Leucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Product Code. 11402707
Click to view available options
Quantity:
25 g
100 g
500 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7517-19-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012494
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
Synonym l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride
PubChem CID 2723695
IUPAC Name methyl (2S)-2-amino-4-methylpentanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC(C)C
Specifications
Melting Point 149°C to 151°C
Beilstein 3595133
Formula Weight 181.66
Percent Purity 99%
Quantity 100 g
Optical Rotation +13° (c=2 in Water)
Sensitivity Hygroscopic
Chemical Name or Material L-Leucine methyl ester hydrochloride