Learn More
L-Leucine, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
Product Code. 15885308
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
Chemical Identifiers
CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2 S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O
Specifications
Melting Point >300°C
Beilstein 1721722
Packaging Plastic Bottle
Formula Weight 131.17
Percent Purity 98.5 to 101.5%
Density 1.293
Quantity 1 kg
Specific Gravity 1.293
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Leucine