missing translation for 'onlineSavingsMsg'
Learn More
L-Lactic Acid (Free Acid), MP Biomedicals™ Product Code.: 11471681

L-Lactic Acid (Free Acid), MP Biomedicals™

Product Code. 11471681 Shop All MP Biomedicals Products
1 g
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy

Product Code. 11471681

Brand: MP Biomedicals 0219022801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00064266
InChI Key JVTAAEKCZFNVCJ-REOHCLBHSA-N
Synonym l-lactic acid, l-+-lactic acid, s-2-hydroxypropanoic acid, s-lactic acid, sarcolactic acid, s-2-hydroxypropionic acid, 2s-2-hydroxypropanoic acid, +-lactic acid, espiritin, tisulac
PubChem CID 107689
ChEBI CHEBI:422
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Color White
Formula Weight 90.08
Density 1190 to 1250kg/mL
Quantity 1 g
Physical Form Solid
Chemical Name or Material L-Lactic Acid, Free Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Lactic Acid (Free Acid), MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.