missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Isoleucine, 99%, MP Biomedicals
348.74 NOK - 4006.80 NOK
Chemical Identifiers
CAS | 73-32-5 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00064222 MFCD00004268 |
InChI Key | AGPKZVBTJJNPAG-WHFBIAKZSA-N |
Synonym | l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile |
PubChem CID | 6306 |
ChEBI | CHEBI:17191 |
IUPAC Name | (2S,3S)-2-amino-3-methylpentanoic acid |
SMILES | CC[C@H](C)[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491881
|
MP Biomedicals
0219468901 |
1 g |
411.28 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401891
|
MP Biomedicals
0219468905 |
5 g |
348.74 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11411891
|
MP Biomedicals
0219468925 |
25 g |
1107.70 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11421891
|
MP Biomedicals
0219468980 |
100 g |
4006.80 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
73-32-5 | |
131.18 | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
6306 | |
(2S,3S)-2-amino-3-methylpentanoic acid |
C6H13NO2 | |
MFCD00064222 MFCD00004268 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
CHEBI:17191 | |
CC[C@H](C)[C@H](N)C(O)=O |
Specifications
73-32-5 | |
MFCD00064222 MFCD00004268 | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
(2S,3S)-2-amino-3-methylpentanoic acid | |
6306 | |
131.2 | |
Crystalline |
C6H13NO2 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
CC[C@H](C)[C@H](N)C(O)=O | |
131.18 | |
CHEBI:17191 | |
Cell Culture Reagent | |
L-Isoleucine, 99% |
Safety and Handling
Recommended Storage : Room temperature