missing translation for 'onlineSavingsMsg'
Learn More
L-Histidine Monohydrochloride Monohydrate, MP Biomedical Product Code.: 11334429

L-Histidine Monohydrochloride Monohydrate, MP Biomedical

Product Code. 11334429 Shop All MP Biomedicals Products
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 11334429

Brand: MP Biomedicals 0210195780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrateShow MoreShow Less
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Color White
Quantity 100 g
Formula Weight 209.6
Physical Form Powder
Chemical Name or Material L-Histidine Monohydrochloride Monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Histidine Monohydrochloride Monohydrate, MP Biomedical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.