missing translation for 'onlineSavingsMsg'
Learn More
L(+)-Ascorbic Acid, ACS Reagent Grade, Scharlab Product Code.: 11399853

L(+)-Ascorbic Acid, ACS Reagent Grade, Scharlab

Product Code. 11399853
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy

Product Code. 11399853

Brand: Scharlab AC05151000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the synthesis of organic products, for pharmaceutical use, as an antioxidant (in food industry) and in biochemistry.

  • Synonyms: Vitamin C, 3-Oxo-L-gulonic acid-g-lactone
  • Molecular Formula: C6H8O6
  • Molar Mass: 176.13g/mol
  • CAS: 50-81-7
  • EIN ECS No.: 200-066-2
  • Ignition Temperature: 380°C

Specifications

CAS 50-81-7
Melting Point 190°C to 192°C (decomposition)
Ignition Residue 0.05% max
Loss on Drying 0.1% max (105°C)
Molecular Formula C6H8O6
MDL Number MFCD00064328
Quantity 1 kg
Specific Rotation 20.5–21.5° ([a]20°/D, c=10, water)
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
Solubility Information In water: 330g/L at 24°C
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular Weight (g/mol) 176.12
PubChem CID 54670067
ChEBI CHEBI:29073
Formula Weight 176.13g/mol
Percent Purity 99.7% min (iodometric)
Grade ACS Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L(+)-Ascorbic Acid, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.