missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)Arabinose, Extra Pure, SLR, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
CAS | 5328-37-0 |
Melting Point | 155°C |
Color | White |
Packaging | Plastic powder jar |
Molecular Formula | C5H10O5 |
MDL Number | 135866 |
Quantity | 25 g |
InChI Key | PYMYPHUHKUWMLA-UHFFFAOYNA-N |
SMILES | OCC(O)C(O)C(O)C=O |
IUPAC Name | 2,3,4,5-tetrahydroxypentanal |
Show More |
Safety and Handling
Product Identifier
- L(+)Arabinose
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L(+)Arabinose, Extra Pure, SLR, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction