missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, MP Biomedicals™

Plays a key role in the glucose-alanine cycle between muscle tissue and the liver. L-Alanine, MP Biomedicals is a nonessential amino acid. In amino acid-degrading tissues, such as muscle, amino groups are pooled as glutamate by transamination reactions.

805.60 NOK - 4197.60 NOK

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11422970
View Documents Promotion Details
MP Biomedicals
02100287.1
100 g
805.60 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
11432970
View Documents Promotion Details
MP Biomedicals
02100287.5
500 g
4197.60 NOK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The amino group of glutamate is transferred to pyruvate via alanine aminotransferase, forming alanine and α-ketoglutarate. The alanine is passed into the blood and transported to the liver. This reaction is reversed in the liver, where pyruvate can be used in gluconeogenesis to form glucose, which may return to other tissues through the circulatory system. Increased alanine levels correlate with higher blood pressure, energy intake, cholesterol levels, and body mass index.

Alanine has been used as an amino donor in a study of the conversion of ketomethiobutyrate to methionine in several species of Bacillus. L-Alanine has been shown to enhance the rates of germination of Bacillus subtilis spores.

Specifications

Chemical Identifiers

56-41-7
89.09
QNAYBMKLOCPYGJ-REOHCLBHSA-N
5950
(2S)-2-aminopropanoic acid
C3H7NO2
MFCD00064410
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
CHEBI:16977
C[C@H](N)C(O)=O

Specifications

56-41-7
MFCD00064410
Freely soluble in water; sparingly soluble in alcohol; insoluble in ether.
C[C@H](N)C(O)=O
89.09
CHEBI:16977
≥99%
C3H7NO2
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
QNAYBMKLOCPYGJ-REOHCLBHSA-N
(2S)-2-aminopropanoic acid
5950
89.09
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature (15–30°C)

SDS
Documents

Documents

Product Certifications