missing translation for 'onlineSavingsMsg'
Learn More

L-Alanine, 99%, MP Biomedicals™

A nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions

362.52 NOK - 30024.50 NOK

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11412970
View Documents Promotion Details
MP Biomedicals
02100287.3
25 g
362.52 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
11442970
View Documents Promotion Details
MP Biomedicals
0210028701
1 kg
6312.30 NOK
1kg
Please to purchase this item. Need a web account? Register with us today!
11452970
View Documents Promotion Details
MP Biomedicals
0210028705
5 kg
30024.50 NOK
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

56-41-7
89.09
QNAYBMKLOCPYGJ-REOHCLBHSA-N
5950
(2S)-2-aminopropanoic acid
C3H7NO2
MFCD00064410
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
CHEBI:16977
C[C@H](N)C(O)=O

Specifications

56-41-7
0.99
MFCD00064410
QNAYBMKLOCPYGJ-REOHCLBHSA-N
(2S)-2-aminopropanoic acid
5950
89.1
Powder
White
C3H7NO2
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
C[C@H](N)C(O)=O
89.09
CHEBI:16977
1.40g/mL
L-Alanine
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-273-8

SDS
Documents

Documents

Product Certifications