Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)
Designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter
23770.00 NOK - 23900.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
11867513
|
DWK Life Sciences
953825-0090 |
1 L |
23770.00 NOK
Each |
Estimated Shipment: 11-03-2026
Log in to see stock. |
||||||
11877513
|
DWK Life Sciences
953835-0090 |
2 L |
23900.00 NOK
Each |
Estimated Shipment: 11-03-2026
Log in to see stock. |
||||||
Description
This unit is also recommended for filtering large volumes of HPLC solvents.
- The ground glass connection eliminates the possibility of phthalate contamination that can occur when using silicone or neoprene stoppers
- Each apparatus is supplied with 1000mL funnel; 90mm, 40-60 micron porosity, fritted glass support base; anodized aluminum clamp; and filtration flask
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Glass | |
Microfiltration Assembly |
Product Title
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)