Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Reusable Threaded Centrifuge Tubes
Heavy wall withstands high centrifuge speeds
Brand: DWK Life Sciences 45212-35
2153.58 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Heavy wall withstands high centrifuge speeds
Ideal for separating neutralized sediment for culturing, from specimens of sputum in clinical work and microbiological applications.
- Marking spot is sandblasted
- Caps are not attached
- Supplied with autoclavable caps with rubber liners
- GPI thread finish is 24mm-410
Specifications
USP, ASTM E 438 | |
Borosilicate Glass | |
Rubber | |
Autoclavable | |
100 mm | |
Round | |
29 mm | |
3550 x g |
35 mL | |
Useful in separating neutralized sediment for culturing from specimens of sputum in clinical work and microbiological applications | |
24-410 | |
Round | |
1.15 oz. | |
Marking Spot | |
Centrifuge Tube |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%