missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20162-1-AP-150UL
This item is not returnable.
View return policy
Description
KLHL29 (Kelch-like protein 29), also known as KBTBD9, is a 655 amino acid protein that is related to the Drosophila Kelch protein. Mutations affecting Kelch function result in failure of Kelch to associate with the ring canals and subsequent female sterility. Human KLHL29 contains six kelch repeats and one BTB (POZ) domain. The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KLHL29 exists as two alternatively spliced isoforms which are encoded by a gene that maps to human chromosome 2p24.1.Specifications
KLHL29 | |
Polyclonal | |
Unconjugated | |
KLHL29 | |
KBTBD9, kelch like 29 (Drosophila), Kelch like protein 29, KIAA1921, KLHL29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114818, 208439, 298867 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80T74, Q96CT2 | |
Klhl29 | |
KLHL29 Fusion Protein Ag14087 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction