Learn More
KLHL14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16693-1-AP-20UL
 View more versions of this product
Product Code. 16896404
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
KLHL14 is a member of the KLHL family.
Specifications
Specifications
KLHL14
Polyclonal
Unconjugated
KLHL14
kelch like 14 (Drosophila), Kelch like protein 14, KIAA1384, KLHL14
Rabbit
Antigen Affinity Chromatography
RUO
225266, 364823, 57565
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q69ZK5, Q9P2G3
Klhl14
KLHL14 Fusion Protein Ag10065
20 μL
Primary
Human, Rat, Mouse