Learn More
Klever Klever Excel Narrow Cut
Recessed blade of the Klever Excel Narrow Cut reduces cut injuries and damaged goods.
2691.05 NOK - 3752.40 NOK
Specifications
Product Type | Klever Excel Narrow Cutter |
---|---|
Disposable | Yes |
Sterility | Non-sterile |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17758987
|
Klever
KCJ-4-20B |
Blue |
3752.40 NOK
Pack of 50 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
17768987
|
Klever
KCJ-4-20G |
Orange |
3752.40 NOK
Pack of 50 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
17778987
|
Klever
KCJ-4-20R |
Red |
2691.05 NOK
Pack of 50 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
17788987
|
Klever
KCJ-4-20Y |
Yellow |
2691.05 NOK
Pack of 50 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
Description
- Cuts boxes, banding, film, tape, and thousands more applications.
- Multiple position handle; two positions for control and reach.
- Available in both narrow and wide heads with durable, high quality carbon steel blade.
- Innovative plastic tape splitter.
- Advanced Plastic Polymers.
- Highest Safety Level.
- Disposable.
Specifications
Klever Excel Narrow Cutter | |
Yes | |
Non-sterile |