missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G80 Neoprene/Latex Chemical Resistant Gloves

Product Code. 15356738
Click to view available options
Size:
Large
Medium
Small
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

G80 gloves have a natural rubber latex construction covered in neoprene to give a high resistance to acids, alkalis, oils and solvents. Ideal for removing paint, sealants and residues from a surface or degreasing machinery with a strong solvent.

  • Neoprene over natural rubber latex construction with cotton flock lining for comfort and perspiration absorption
  • High resistance to acids, oils, solvents and alkalis
  • Meets the EN 374-AKL standard and has a high level of dexterity, per EN420:2003 standard
  • Not suitable for those with Type 1 latex allergies

Specifications

Certifications/Compliance (Europe) EN 420:2003, EN 374
Applications Industrial
Color Blue, Yellow
Latex Free No
Material Neoprene, Latex
Product Type Chemical-resistant Gloves
Size Medium
Thickness (English) 27.5 mil
Thickness (Metric) 0.69 mm
EN ISO 374-1 CHEMICAL LETTER CODES AKL
Glove Cuff Style Straight
Grip Design Textured Fingertips
Liner Material Cotton Flock
Lining Lined
Model 38742
Series Kleenguard™ G80
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.