Learn More
Lenz Laborglasintrumente Kjeldahl Flasks
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03051928
 View more versions of this product
Product Code. 10570112
  • 4680.00 NOK / Pack of 10
Only null left
Estimated Shipment: 22-08-2025
Description
  • For the determination of nitrogen in organic compounds
  • With center neck
  • With conical joint and inscription label
  • Material: DURAN™
Specifications
Specifications
50 mL
NS 19/26
10/Pk.
Borosilicate Glass (3.3)
51 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Kjeldahl Flasks > 50 mL