Learn More
Lenz Laborglasintrumente Kjeldahl Flasks
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03051937
 View more versions of this product
Product Code. 10264621
  • 4875.00 NOK / Pack of 10
Only null left
Estimated Shipment: 15-08-2025
Description
  • For the determination of nitrogen in organic compounds
  • With center neck
  • With conical joint and inscription label
  • Material: DURAN™
Specifications
Specifications
100 mL
NS 19/26
10/Pk.
Borosilicate Glass (3.3)
60 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Kjeldahl Flasks > 100 mL