Learn More
Thermo Scientific™ KingFisher™ Systems Consumables
Optimize performance using Thermo Scientific™ KingFisher™Systems Consumables, ideal for magnetic particle processing.
676.00 NOK - 2975.00 NOK
Specifications
For Use With (Application) | Magnetic particle processing |
---|---|
For Use With (Equipment) | KingFisher System |
Quantity | 50/Cs. |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11520625
|
Thermo Scientific™
97002070 |
KingFisher tip comb |
676.00 NOK
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12673116
|
Thermo Scientific™
97002080 |
KingFisher plate 100 ÎĽL |
2975.00 NOK
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12683116
|
Thermo Scientific™
97002084 |
KingFisher plate 200 ÎĽL |
2935.00 NOK
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
- Specifically designed for use with the KingFisher System
- Ideal for magnetic particle processing because of their low binding affinity for biomolecules
- The specially-designed, disposable tip combs and microplates are made of polypropylene
- This enables excellent recovery of magnetic beads
Specifications
Magnetic particle processing | |
KingFisher System | |
50/Cs. |