Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Brand:  Thermo Scientific™ 95040460B
2360.11 NOK valid until 2025-06-21
Use promo code "24112" to get your promotional price.
 View more versions of this product
Product Code. 17130069
  • 3540.00 NOK / Pack of 50
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile 96 Deep Well Plate
KingFisher 96 Deep-Well Plate, Sterile, Barcoded
KingFisher Apex
50/Cs.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number24154B0 Certificate TypeCertificate of Compliance Date04/10/2024 Catalog Number95040460B
Lot Number24135B0 Certificate TypeCertificate of Compliance Date29/08/2024 Catalog Number95040460B
Lot Number21250B0 Certificate TypeCertificate of Compliance Date07/10/2021 Catalog Number95040460B
Lot Number21249B0 Certificate TypeCertificate of Compliance Date21/09/2021 Catalog Number95040460B
Lot Number21245B0 Certificate TypeCertificate of Compliance Date21/09/2021 Catalog Number95040460B
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format >