Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Brand:  Thermo Scientific™ 97003520
450.68 NOK valid until 2025-06-21
Use promo code "24112" to get your promotional price.
 View more versions of this product
Product Code. 12633116
  • 676.00 NOK / Pack of 40
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Elution Strip
KingFisher elution strip for 12 pin magnet (for Duo Prime only)
KingFisher Duo Prime only
40 Pc.
Documents
Product Bulletins
Specification Sheets
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2417810 Certificate TypeCertificate of Compliance Date04/10/2024 Catalog Number97003520
Lot Number2411510 Certificate TypeCertificate of Compliance Date28/08/2024 Catalog Number97003520
Lot Number2411210 Certificate TypeCertificate of Compliance Date28/08/2024 Catalog Number97003520
Lot Number2411010 Certificate TypeCertificate of Compliance Date27/06/2024 Catalog Number97003520
Lot Number2411310 Certificate TypeCertificate of Compliance Date03/06/2024 Catalog Number97003520
Promotions
Promotions
  • Invitrogen , ABI Applied Biosystems , Thermo Scientific , Affymetrix eBioscience
    Exclusive savings to help your research thrive.
    Promo Code: 24108
    Expires2025-06-27
    New Offer
    Save Now
  • Multiple Manufacturers
    Exclusive savings, limited-time offer.
    Promo Code: 24112
    Expires2025-06-20
    New Offer
    Save Now
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format > KingFisher Duo elution strip