Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Brand: Thermo Scientific™ 95040460
1970.09 NOK valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Specifications
Sterile 96 Deep Well Plate | |
96 Samples | |
KF Duo Prime, Flex and Presto |
96 Deep-well Plate | |
KingFisher 96 deep-well plate, sterile (for Duo Prime, Flex and Presto) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
20280B0 | Certificate of Compliance | 13/11/2024 | 95040460 |
24234B0 | Certificate of Compliance | 29/10/2024 | 95040460 |
24152B0 | Certificate of Compliance | 04/10/2024 | 95040460 |
24102B0 | Certificate of Compliance | 04/10/2024 | 95040460 |
24100B0 | Certificate of Compliance | 04/10/2024 | 95040460 |
Promotions
-
Multiple Manufacturers
Get 3 for 2
Exclusive savings, limited-time offer.Promo Code: 24112Expires: 2025-06-20
Days
7Hours
18Minutes
10Seconds
47New Offer Save Now