Learn More
DWK Life Sciences Kimble™ Kontes™ Flow Adapter Replacement Components for CHROMAFLEX™ Columns
Replacement HDPE Bed Supports for CHROMAFLEX™ Flow Adapters
1630.00 NOK
Specifications
Material HDPE
View More Specs
Products 1
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
11843063
View Documents
DWK Life Sciences
420811-2020
2.5 cm
1630.00 NOK
Pack of 10
Estimated Shipment: 04-04-2025
Description
  • HDPE Bed Support
  • 20μm HDPE Bed Support
  • 1.0cm, 2.5cm and 4.8cm options
  • Case of 10
Specifications
Specifications
HDPE