Learn More
DWK Life Sciences Kimble™ Conical-Bottom Glass Centrifuge Tubes: 5mL Capacity
Borosilicate Glass
Brand: DWK Life Sciences 73790-5
This item is not returnable.
View return policy
Description
Borosilicate Glass
- Capacity: 5mL
- O.D. x L: 13 x 110mm
Specifications
USP, ASTM E 438 | |
Single-use | |
Conical | |
Conical | |
13 mm | |
2980 x g |
5 mL | |
Borosilicate Glass | |
110 mm | |
Not Included | |
Centrifuge Tube |
KIM001 Kimble Chase 08 09 Catalog
File size: 57.1 MB
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Conical-Bottom Glass Centrifuge Tubes: 5mL Capacity > Capacity: 5mL; O.D. x L: 13 x 110mm; Without caps