Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials w/Attached PTFE-Faced/White Rubber Lined Closures
Attached closed top black phenolic closures
3085.00 NOK - 6650.00 NOK
Specifications
Material (Liner) | PTFE/Rubber |
---|---|
Color | Amber |
Material | Borosilicate Glass |
Thread Style | Screw |
Closure Material | Phenolic |
Product Code | Brand | Volume (Metric) | Height (Metric) | Diameter (Metric) Outer | Volume (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Height (Metric) | Diameter (Metric) Outer | Volume (English) | Price | Quantity & Availability | |||
11852973
|
DWK Life Sciences
60911B-1 |
4 mL | 45 mm | 15 mm | 1 dr. |
3845.00 NOK
Pack of 144 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
11833003
|
DWK Life Sciences
60911B-1528 |
2 mL | 28 mm | 15 mm | 0.5 dr. |
4625.00 NOK
Pack of 200 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
11833023
|
DWK Life Sciences
60911B12 |
1.8 mL | 35 mm | 12 mm | 0.5 dr. |
6650.00 NOK
Pack of 288 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
11843003
|
DWK Life Sciences
60911B1738 |
4 mL | 38 mm | 17 mm | 1 dr. |
4915.00 NOK
Pack of 200 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
11853003
|
DWK Life Sciences
60911B1940 |
5.5 mL | 40 mm | 19 mm | 1.49 dr. |
4685.00 NOK
Pack of 200 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
11872953
|
DWK Life Sciences
60911B-5 |
20 mL | 57 mm | 28 mm | 5.4 dr. |
3085.00 NOK
Pack of 72 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
11862953
|
DWK Life Sciences
60911B-10 |
40 mL | 95 mm | 28 mm | 10.82 dr. |
4035.00 NOK
Pack of 72 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
Description
- Manufactured from amber 51 expansion borosilicate glass
- Closures with PTFE-faced/white rubber liners or white rubber liners
- Vials packaged in corrugated trays with partitions
Specifications
PTFE/Rubber | |
Borosilicate Glass | |
Phenolic | |
USP Type I |
Amber | |
Screw | |
Screw Cap (Closed-top) |
Product Title
DWK Life Sciences Kimble™ AmberScrew Thread Vials w/Attached PTFE-Faced/White Rubber Lined Closures