Learn More
DWK Life Sciences Kimble™ 51 Expansion Borosilicate Glass Melting Point Capillaries with Smaller I.D.
51 expansion borosilicate glass
Brand:  DWK Life Sciences 34502-99
 View more versions of this product
Product Code. 11873063
  • 2965.00 NOK / Pack of 20
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Description
  • I.D.: 0.8 to 1.1mm
  • 100 capillaries per vial
Compliance
Comply with U.S. Pharmacopeia XXI.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
Melting Point Capillary (Both Ends Open)
0.8 to 1.1 mm
100 mm
0.25 mm
USP type I, ASTM E438
3.93 in.