missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KG 5
PDGFRβ, B-Raf, c-Raf, FLT3 and KIT inhibitor
1815.00 NOK - 6990.00 NOK
Specifications
Chemical Name or Material | 2-(Methylthio)-6-[4-[5-[[3-(trifluoromethyl)phenyl]amino]-1H-1,2,4-triazol-3-yl]phenoxy]-4-pyrimidinamine |
---|---|
CAS | 877874-85-6 |
Target | Raf Kinase Inhibitors |
Molecular Formula | C20H16F3N7OS |
Formula Weight | 459.45 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780518
|
Tocris Bioscience™
5260/10 |
10 mg |
1815.00 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18780789
|
Tocris Bioscience™
5260/50 |
50 mg |
6990.00 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGFRβ and B-Raf allosteric inhibitor. Also inhibits c-Raf, FLT3 and KIT. Disrupts blood vessel formation in zebrafish embryos and inhibits angiogenesis in a mouse angiogenesis model. Inhibits growth of renal and pancreatic tumors in mice. Also arrests cells in prometaphase. Orally available.Specifications
2-(Methylthio)-6-[4-[5-[[3-(trifluoromethyl)phenyl]amino]-1H-1,2,4-triazol-3-yl]phenoxy]-4-pyrimidinamine | |
Raf Kinase Inhibitors | |
459.45 |
877874-85-6 | |
C20H16F3N7OS | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ KG 5