Learn More
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Brand:  Thermo Scientific™ 97002610B
6188.00 NOK valid until 2025-06-30
Use promo code "24108" to get your promotional price.
 View more versions of this product
Product Code. 17190069
  • 7280.00 NOK / Pack of 50
Nurture Your Progress
Use promo code 24108 at checkout
View Available Promotions
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 DW tip comb, plate, barcode
KingFisher Apex
50/Cs.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2425010 Certificate TypeCertificate of Compliance Date21/10/2024 Catalog Number97002610B
Lot Number2424610 Certificate TypeCertificate of Compliance Date10/10/2024 Catalog Number97002610B
Lot Number2419910 Certificate TypeCertificate of Compliance Date04/10/2024 Catalog Number97002610B
Lot Number2419810 Certificate TypeCertificate of Compliance Date04/10/2024 Catalog Number97002610B
Lot Number2417110 Certificate TypeCertificate of Compliance Date04/10/2024 Catalog Number97002610B
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format > KingFisher Apex 24 DW tip comb, plate, barcode