Learn More
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Brand:  Thermo Scientific™ 97003510
1436.50 NOK valid until 2025-04-18
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 12643116
  • 1726.00 NOK / Pack of 48
Explore more special offers
Description
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plate and Comb
KingFisher Duo Prime 6-tip comb and 24 deep-well plate (12 pieces of 24 deep-well plates, each including 4 tip combs)(for Duo Prime only)
For Use With KingFisher Duo Prime only
48 Pc.
Documents
Product Bulletins
Specification Sheets
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2407410 Certificate TypeCertificate of Compliance Date15/08/2024 Catalog Number97003510
Lot Number2407610 Certificate TypeCertificate of Compliance Date25/06/2024 Catalog Number97003510
Lot Number2407010 Certificate TypeCertificate of Compliance Date19/06/2024 Catalog Number97003510
Lot Number2223510 Certificate TypeCertificate of Compliance Date31/08/2022 Catalog Number97003510
Lot Number2223110 Certificate TypeCertificate of Compliance Date31/08/2022 Catalog Number97003510
Product Title
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format > Nucleic acid purification system; Use with KingFisher duo 6-tip combs and 24 deepwell plate