Learn More
KEL Mouse anti-Human, Unconjugated, Clone: 1C8B6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67393-1-IG-20UL
 View more versions of this product
Product Code. 16859995
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The KEL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KEL. This antibody recognizes human antigen. The KEL antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
KEL
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1C8B6
Unconjugated
20 μL
3792
IgG2a