Learn More
DWK Life Sciences KECK™ Screw Cap for Adapter KA, with flat EPDM sealing 12 mm
Shop All DWK Life Sciences Products
Click to view available options
Thread Size:
GL 14
RD 14
Unit Size:
Pack of 100
Description
The screw cap as spare part for KECK™ Adapter KA is available for both thread types GL 14 and RD 14.
- With flat gasket EPDM 12 mm
- KECK™ Screw cap for adapter KA available for threads GL 14 and RD 14
Specifications
Specifications
| Color | Yellow |
| Description | Not Autoclavable |
| For Use With (Application) | Connection of a hose piece with a threaded cap |
| Quantity | 100/Pk. |
| Thread Size | GL 14 |