missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNQ3 Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19966-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody recognizes the C-term of KCNQ3.
KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KCNQs (KCNQ1-5) found in the central nervous system and KCNQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.Specifications
KCNQ3 | |
Polyclonal | |
Unconjugated | |
KCNQ3 | |
BFNC2, EBN2, KCNQ3, KQT like 3, KV7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100157152, 3786 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43525 | |
KCNQ3 | |
Peptide | |
20 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG |