missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNJ1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20953-1-AP-20UL
This item is not returnable.
View return policy
Description
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. It is activated by internal ATP and probably plays an important role in potassium homeostasis. The encoded protein has a greater tendency to allow potassium to flow into a cell rather than out of a cell. Mutations in this gene have been associated with antenatal Bartter syndrome, which is characterized by salt wasting, hypokalemic alkalosis, hypercalciuria, and low blood pressure. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
KCNJ1 | |
Polyclonal | |
Unconjugated | |
KCNJ1 | |
KCNJ1, KIR1.1, ROMK, ROMK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24521, 3758, 56379 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88335, P35560, P48048 | |
KCNJ1 | |
KCNJ1 Fusion Protein Ag15100 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction