Learn More
KATNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17560-1-AP-150UL
 View more versions of this product
Product Code. 16847474
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.
Specifications
Specifications
KATNA1
Polyclonal
Unconjugated
KATNA1
Katanin p60 subunit A1, KATNA1, p60 katanin
Rabbit
Antigen Affinity Chromatography
RUO
11104, 23924, 292464
-20°C
Liquid
Immunoprecipitation, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75449, Q6E0V2, Q9WV86
Katna1
KATNA1 Fusion Protein Ag11753
150 μL
Primary
Human, Mouse, Rat