Learn More
KATNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17560-1-AP-150UL
 View more versions of this product
Product Code. 16847474
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.
Specifications
Specifications
KATNA1
Polyclonal
Unconjugated
KATNA1
Katanin p60 subunit A1, KATNA1, p60 katanin
Rabbit
Antigen Affinity Chromatography
RUO
11104, 23924, 292464
-20°C
Liquid
Immunoprecipitation, Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75449, Q6E0V2, Q9WV86
Katna1
KATNA1 Fusion Protein Ag11753
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KATNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated