Learn More
Robin™ Kahn Complete Shaker
Versatile shaker for mixing or agitating ingredients and solutions in flasks, bottles, beakers, and Kahn racks. Robin™ Kahn Complete Shaker is available in different shaker speed ranges.
Brand: Robin™ PB021
This item is not returnable.
View return policy
Description
- Mixes or agitates ingredients and solutions in a variety of containers including, but not limited to, the following:
- Flasks
- Bottles
- Beakers
- Kahn racks
Specifications
6.5 kg | |
230 V, 50 Hz, 25 W | |
50 Hz | |
250 x 360 x 130 mm | |
360 mm |
40 to 120 rpm | |
25 W | |
250 mm | |
130 mm | |
Kline Stirrer Complete 40-120T |
Product Content Correction
Product Title
Robin™ Kahn Complete Shaker > Speed Range: 40 to 120RPM/min.