Learn More
uvex K4 Ear Muffs
The Uvex K protective ear muffs offer an impressive combination of high levels of protection combined with great design.
Brand: uvex 2600004
327.13 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Soft surface
- Easy to mould
- Comfortable to wear
- Low weight
Specifications
EN 352-1 | |
No | |
Reusable | |
Black | |
Over-the-head |
32 dB | |
Ear Muffs | |
Yes | |
No | |
No |