Learn More
SI Analytics KF Drying Oven
Robust, reliable, ready-to-use drying oven that is ideal for water determination according to Karl Fischer.
Brand: SI Analytics 285214721
This item is not returnable.
View return policy
Description
- Drying oven TO 52, type: TZ1052, max. Temperature + 200 ° C, 230 ° C
- TZ 1052 Drying oven for water determination according to Karl-Fischer, 230 V
- Used or water analysis
Specifications
KF Drying Oven | |
Titrator Accessories | |
230 V |
Product Content Correction
Product Title
SI Analytics KF Drying Oven > Voltage: 230 V; Model TZ1052