Learn More
uvex K-Basic Extra Aramid Fiber Cut/Heat Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
3
8
10
12
Description
- Course-knit aramid fiber gloves for applications that involve both thermal and mechanical risks
- A high level of thermal insulation when in direct contact with warm/hot objects
- Aramid fiber construction provides resistance against cuts
- Cotton liner on the inside increases user comfort
- Ideal for light to medium metalworking, glass handling, and automotive industries
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Certifications/Compliance | EN 388:2016 (2 4 4 2 D) and EN 407:2004 (X 2 X X X X) |
Coating Material | Uncoated |
Color | Yellow |
Latex Free | Yes |
Material | Aramid Fiber |
Product Type | Heat-resistant Gloves |
Size | 3 |
Style | Full-hand Mitt |
Temperature (Metric) Max | 250°C |
Liner Material | Cotton |
Show More |
Product Content Correction
Product Title
uvex K-Basic Extra Aramid Fiber Cut/Heat Protection Gloves >