Learn More
Justrite™ Nonmetallic Self-Close Corrosive Safety Containers
Designed to resist acids and caustics
895.00 NOK - 2350.00 NOK
Specifications
Product Type | Stainless Steel Safety Can |
---|---|
Color | White |
Material | Polyethylene |
Includes | Spring Close |
Shape | Oval |
Product Code | Brand | Capacity (English) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Price | Quantity & Availability | ||||||
11983669
|
Justrite™
12260 |
2.5 gal. | N/A | |||||||
10451165
|
Justrite™
12161 |
1 gal. |
895.00 NOK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
11903679
|
Justrite™
12762 |
2 gal. | N/A | |||||||
11913679
|
Justrite™
12765 |
5 gal. |
2350.00 NOK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
Description
Designed to resist acids and caustics
- Safely holds nonflammable corrosives
- Translucent white HDPE construction resists dents, punctures, and chemicals
- Marked in liters, quarts, and gallons for accurate filling and dispensing
- Flame arrester-free spout allows quick pouring; spring-close cap provides a tight seal
- Wide mouth on Cat. Nos. 17-985-89B, -89C, and -89D makes filling and pouring easier
†Oval-shaped.
Specifications
Stainless Steel Safety Can | |
Polyethylene | |
Oval |
White | |
Spring Close |
Product Title
Justrite™ Nonmetallic Self-Close Corrosive Safety Containers