Learn More
Justrite™ Horizontal and Vertical Drum Vents
For controlled release of flammable vapors; prevent explosions caused by pressure buildup
Marca: Justrite™ 08005
813.17 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Descrizione
- Choose brass, stainless steel or nonmetallic vent
- For petroleum-based liquids, use non-metallic or brass vents
- For chlorinated solvents, use nonmetallic vent with stainless-steel flame arrester
- For corrosives and general solvents, use stainless steel vents
Vertical Drum Vent:
- Has operating valve that opens automatically to prevent pressure buildup inside drum
Horizontal Drum Vent
- Has operating valve that can be opened manually to relieve internal pressure
Order Info
All vents fit 2 in. (5.1cm) bung. Cast Iron EL Fitting for use with 17-186A and 17-186B also available; see cat. no. 19018555.
Certifications
FM approved.
Specifica
FM | |
Opens at 5 psig Internal Pressure | |
Vertical Drums | |
Operating Valve, Plated Steel Flame Arrester |
Vertical Drum Vent | |
Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup | |
Non-metallic Cast Iron |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.